Learn More
Nordvlies WIPEX-STAR wipe for wet cleaning, 80 g/sqm, app. 30x38 cm, solvent resistent, blue
Brand:  Nordvlies 2195
WIPEX-STAR wipe for wet cleaning, 80 g/sqm, app. 30x38 cm, solvent resistent, blue
Product Code. 15770947
  • ONLINE EXCLUSIVE PRICE
    95.35€
    80.40€ / 1 roll
    Save 14.95€
    15% Off
Only null left
Estimated Shipment: 15-01-2026