Learn More
Nordvlies WIPEX-FULLPOWER wipes, 57 g/sqm, app. 40x38 cm, solvent resistent, white
Brand:  Nordvlies 240540
WIPEX-FULLPOWER wipes, 57 g/sqm, app. 40x38 cm, solvent resistent, white
Product Code. 15780957
  • ONLINE EXCLUSIVE PRICE
    120.00€
    101.47€ / 1 roll
    Save 18.53€
    15% Off
Only null left
Estimated Shipment: 23-01-2026