Learn More
Nordvlies WIPEX-FULLPOWER wipes, 57 g/sqm, app. 34x38 cm, z-folded, solvent resistent, white
Brand:  Nordvlies 140540
WIPEX-FULLPOWER wipes, 57 g/sqm, app. 34x38 cm, z-folded, solvent resistent, white
Product Code. 15730927
  • ONLINE EXCLUSIVE PRICE
    124.00€
    85.28€ / Pack of 500
    Save 38.72€
    31% Off
Only null left
Estimated Shipment: 20-01-2026