Description
- PVC coating repels water, oil and grease and remains flexible at low temperatures
- Foam insulation keeps hands warm
- Excellent grip for safety when handling heavy-duty objects
- Curved fingers, wing thumb and soft jersey cotton lining offers superior comfort
- Keeps hands warm, dry and protected from abrasions
Specifications
Specifications
| Certifications/Compliance | Standards CATEGORY II EN 388 : 3121, EN 511 : 111, EN 1149, ANSI |
| Cuff Material | PVC |
| Coating Material | PVC |
| Color | Beige |
| Cut Resistance Rating | Level 1 |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | PVC |
| Powdered | Powder Free |
| Product Type | Heavy-duty Gloves |
| Show More |
Product Title
Individuate un'opportunità di miglioramento?