Learn More
Win 18446, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Win 18446 |
| CAS | 1477-57-2 |
| Molecular Formula | C12 H20 Cl4 N2 O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C12H20Cl4N2O2/c13-9(14)11(19)17-7-5-3-1-2-4-6-8-18-12(20)10(15)16/h9-10H,1-8H2,(H,17,19)(H,18,20) |
| SMILES | ClC(Cl)C(=O)NCCCCCCCCNC(=O)C(Cl)Cl |
| IUPAC Name | 2,2-dichloro-N-[8-[(2,2-dichloroacetyl)amino]octyl]acetamide |
| Molecular Weight (g/mol) | 366.11 |
| Formula Weight | 364.0279 |