Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Shigemi™ Susceptibility Matched NMR Tubes
Shop All SP Scienceware Products
Click to view available options
Model:
BMS-003
BMS-005B
CMS-003
CMS-005B
DMS-003
MMS-003
Diameter (Metric) Outer:
3 mm
5 mm
For Use With (Equipment):
Bruker Spectometers
Universal, All Spectometers
Unit Size:
Each
Description
- Select tube based on solvent used: CDCL3, CD3OD, D2O, DMSO-d6
- Available for Bruker or Universal
Specifications
Specifications
| For Use With (Equipment) | Universal, All Spectometers |
| Length (Metric) | 10 mm (Bottom) |
| Material | Glass (Type 1, Class A) |
| Product Type | Tube Set |
| Diameter (Metric) Outer | 3 mm |
| For Use With (Application) | Deuterium Oxide Matched |
| Model | BMS-003 |