Learn More
Thermo Scientific Chemicals Wig-L-Bug™ Accessories
Product Code. 15402007
Click to view available options
Quantity:
100/Pk.
25/Pk.
5/Pk.
Each
Type:
Vial
Vial Adapter
Unit Size:
Each
Pack of 100
Pack of 25
Pack of 5
Height (English):
2 in.
1 ft.
2 ft.
Product Code. 15402007
Brand: Thermo Scientific Chemicals 044531.KT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use with Digital Wig-L-Bug Mixing/Grinding Mill
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Polystyrene vial with polyethylene slip-on cap; 1 ft. long x 0.5 in. dia.
  • Polystyrene vial with polyethylene slip-on cap; 2 ft. long x 0.5 in. dia.
  • Adapter for 2 in. long, 0.5 in. diameter vials
TRUSTED_SUSTAINABILITY
Specifications
Description Accessory for Digital Wig-L-Bug mixing/grinding mill
Quantity Each
Type Vial Adapter