Learn More
Motic™ Widefield Eyepiece WF15X/13mm
Click to view available options
Magnification Power:
15X
Unit Size:
Each
Description
- Widefield style provides improved field of view and greater eye relief.
- Magnification: 15X.
- Field of view: 13mm.
Specifications
Specifications
| Type | Widefield |
| Description | FOV: 13mm |
| For Use With (Equipment) | ST-39, SFC-11, SMZ-140 Microscopes |
| Magnification Power | 15X |