Learn More
Karl Hecht™ Assistent™ Glass Powder Funnels
Click to view available options
Diameter (Metric)Top:
40mm
60mm
100mm
Unit Size:
Each
Description
- With short, wide stem
- Available in a range of diameters
Specifications
Specifications
| Diameter (Metric)Top | 100mm |
| Material | Glass |
| For Use With (Application) | Laboratory Applications |