Learn More
Hirschmann™ Wide Neck Square Bottle, 100-pack
Product Code. 16457019
Change view
Click to view available options
Capacity (Metric):
20 mL
30 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
1500 mL
Unit Size:
Pack of 100
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
16457019 50 mL Pack of 100
16417019 20 mL Pack of 100
16437019 30 mL Pack of 100
16477019 100 mL Pack of 100
16497019 250 mL Pack of 100
16417029 500 mL Pack of 100
16437029 1000 mL Pack of 100
16457029 1500 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 16457019 Supplier Hirschmann™ Supplier No. 7690675
Only null left
Add to basket
Edge
Add to basket
High quality wide neck square bottle made of HDPE with a screw cap made of LDPE is designed to facilitate easier filling and emptying.
  • Wide neck
  • Square
  • HDPE
  • Starting from size 20 mL and up
  • Natural
  • With screw cap made of LDPE
Specifications
Bottle Surface Transparent
Capacity (Metric) 50 mL
Cleaning Procedure Gas/Chemical/Radiation
Color Natural
Diameter (Metric) Outer Neck 28 mm
Diameter (Metric) Inner Neck 21.5 mm
Material HDPE
Neck ID (Metric) 21.5 mm
No. per Pack 100
Sterility Non-sterile
Thread Size (Metric) GL 28
Closure Color Natural
Content And Storage Store at 5 to 40°C
Closure Included Yes
Shape Square
Height (Metric) 74 mm
Closure Material LDPE
Closure Size GL 28
Diameter (Metric) 35 x 35 mm
Closure Type Screw Cap
Autoclavable Not Autoclavable