Learn More
BRAND™ Wide Neck Graduated Erlenmeyer Flask
Shop All Brand Products
Click to view available options
:
Capacity: 50 mL
Description
- High clarity
- With graduations
- Neck with standard ground socket (Conical joint stopper, PE-LD: NS 34/35 Cat. No. 1444 40, NS 45/40 Cat. No. 1444 45.)
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Polypropylene |
| Neck Style | Wide neck |
| Product Type | Erlenmeyer Flask |
| Graduated | Yes |
| Closure Type | Screw |
| Closure Size | NS 34/35 |