Learn More
BRAND™ Wide Neck Glass Erlenmeyer Flasks
Product Code. p-8772021 Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
300 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Product Code. 10089860
Brand: BRAND™ 92836
With beaded rim and graduations
  • Manufactured from Boro 3.3 glass
  • Wide neck for ease of filling
  • Acc. to DIN EN ISO 24450
  • Provided in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer Neck 50 mm
Capacity (Metric) 250 mL
Material Borosilicate Glass (3.3)
Height (Metric) 140 mm
No. per Pack 10
Diameter (Metric) Outer 85 mm