missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific Bench-Top Keeper™ Biohazard Container
Shop All Heathrow Scientific Products

Click to view available options
Height (Metric):
18 cm
25 cm
Length (Metric):
10 cm
15 cm
Width (Metric):
10 cm
15 cm
Size:
Small
Large
Unit Size:
Pack of 100
Pack of 50
Description
- Made of sturdy paperboard with an internal plastic lining and webbed corners to contain moderate amounts of fluids
- Self-locking tops keep contents safely enclosed
- Small Keepers fold out to 4L x 4W x 7H in. (10 x 10 x 18 cm) and accommodate up to 1600 cc of material
- Large Keepers fold out to 6L x 6W x 10H in. (15 x 15 x 25 cm) and accommodate up to 5000 cc of material
- Autoclavable
- Made in USA
Specifications
Specifications
| Capacity (Metric) | 3 L |
| Capacity (English) | 3000 cc |
| Dimensions (L x W x H) | 15 x 15 x 25 cm (6 x 6 x 10 in.) |
| Disposable | Single-use |
| Height (Metric) | 25 cm |
| Length (Metric) | 15 cm |
| Size | Large |
| Width (Metric) | 15 cm |
| Autoclavable | Autoclavable |
| Includes | Self-locking tops |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction