Learn More
Wheaton™ White Replacement Cap for Urine Tube
Product Code. 16484959
Change view
Click to view available options
Closure Color:
White
Unit Size:
Pack of 5000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Unit Size
16484959 White Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16484959 Supplier Wheaton™ Supplier No. 327225
  • ONLINE EXCLUSIVE PRICE
    424.60€
    297.54€ / Pack of 5000
    Save 127.06€
    29% Off
Estimated Shipment: 11-12-2026
Spare polypropylene cap for primary urine tube.
  • Supports use of Wheaton™ primary urine tubes
  • For use with 10 mL tubes
Specifications
Product Type Cap
Closure Color White
Material Polypropylene
Sterility Non-sterile
Description Replacement cap for primary urine tube, undosed
For Use With (Equipment) 10 mL Urine Tube
No. per Case 5000/Cs.
No. per Pack 1000/Pk.
Storage Requirements Ambient