Learn More
Deutsch Neumann™ White PTFE Tubing
Product Code. 10336924
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
1.5 mm
3 mm
10 mm
Diameter (Metric) Outer:
3 mm
5 mm
12 mm
Unit Size:
5m
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Unit Size
10336924 1 mm 3 mm 5m
10001124 10 mm 12 mm 5m
10421084 1.5 mm 3 mm 5m
10274954 3 mm 5 mm 5m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10336924 Supplier Deutsch Neumann™ Supplier No. 3400103
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
White PTFE Tubing is temperature resistant up to 270°C in the absence of active pressure.
  • Manufactured from polytetrafluorethylene (PTFE)
  • Minimum order length of 5 m
  • Almost totally chemically inert
  • Excellent flexibility and strength, even in low temperatures
Specifications
Certifications/Compliance ISO 9001:2008
Diameter (Metric) Inner 1 mm
Material PTFE
Diameter (Metric) Outer 3 mm
Thickness (Metric) 1 mm
Packaging Quantity 1 Ea.
Color White