Learn More
3M™ White Disposable Visitor Lab Coat, Series 4400
Catalog Number p-8004233
142.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
142.00 EUR valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Offers a barrier against non-hazardous particles and liquid splashes. The Series 4440 Disposable Protective Lab Coat is anti-static treated and can be used in light duty applications.
  • Breathable, lightweight material.
  • Designed for light duty applications.
  • Offers a barrier against non-hazardous particles and liquid splashes.
  • Zip fastener.
  • Anti-static treated.
  • Knitted cuffs.
Type Lab Coat
Size 4X-Large
Material Polypropylene
Color White
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Resistance Non-hazardous Liquids, Dust
Seam Type Serged
Size (English) Chest 46 to 52 in.
Size (Metric) Chest 117 to 132 cm
Certifications/Compliance CE Category 1
For Use With (Application) Non-hazardous liquids and dust
Includes Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
Packaging Type Individual packaging