Learn More
3M™ White Polypropylene/Polyethylene Laminate Film Protective Coveralls
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Gender | Unisex |
| Disposable | Single-use |
| Waist Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 3-panel |
| Sterility | Non-sterile |
| Protection Class | Category III |
| Height (Metric) | 174 to 181 cm |
| Style | Enhanced Gusset |