Learn More
Fisherbrand™ White Polypropylene Caps
Click to view available options
Closure Size:
8-425 mm
13-425 mm
48-400 mm
70-400 mm
Quantity:
144
500
576
1700
Description
- Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
- PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
- Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Product Type | Lined Cap |
Color | White |
Quantity | 576 |
Closure Color | White |
Closure Liner | Pulp/Vinyl |
Closure Size | 48-400 mm |
Closure Material | Polypropylene |