Learn More
3M™ White Polyethylene Laminate Category I Protective Coveralls
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | 4X-Large/5X-Large |
| Gender | Unisex |
| Disposable | Single-use |
| Waist Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 2-panel |
| Sterility | Non-sterile |
| Protection Class | Category I |
| Show More |