Learn More
Socorex™ White Capillary Tubes
Product Code. p-8010143
Click to view available options
Volume (Metric):
1 to 5 μL
5 to 25 μL
Unit Size:
Pack of 200
Product Code. 10548442
Brand: Socorex™ 341.005
Only null left
Add to basket
Add to basket
Make a connection to laboratory pipettes with Socorex™ White Capillary Tubes. Each tube is supplied in a practical box with a lid.
  • White
  • Replacement glass capillaries
  • For Acura™ 846 positive displacement pipettes
  • Color coded to match corresponding pipette
Specifications
Volume (Metric) 1 to 5 μL
Sterility Sterile
Color White
Quantity 200/Pk.
Product Type Capillary Tube
Disposable Single-use
For Use With (Equipment) Acura 846 Positive Displacement Pipette
Material Glass