missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies WHI-P131

Product Code. p-7217679
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 17130242

Brand: STEMCELL Technologies 73542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

JAK/STAT pathway inhibitor; Inhibits JAK3

  • WHI-P131 is an inhibitor of Janus kinase 3 (JAK3) with IC50 values of 9 and 78 μM against human and mouse proteins, respectively (Sudbeck et al.) It has also been reported to show significant inhibition of other kinases, including epidermal growth factor receptor (EGFR) in the nanomolar range (Changelian et al.; Uckun et al.) No significant inhibition of JAK1 or JAK2 has been observed (Sudbeck et al.)
  • Induces apoptosis and cell death in human glioblastoma cell lines U373 and U87 (Narla et al.)
  • Inhibits lipopolysaccharide (LPS)-induced nitric oxide synthase expression and nitric oxide production in macrophages (Sareila et al.)
  • Inhibits proliferation of neural stem cell-containing neurospheres at lower concentrations than the concentration required to inhibit proliferation of astrocyte cultures (Diamandis et al.)

Specifications

CAS 202475-60-3
Synonym Janex-1
Quantity 1 mg
Molecular Formula C16H15N3O3
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.