Learn More
DWK Life Sciences Wheaton™ V Vial™ with Crimp Top Finish; no Closures
Product Code. 12663355 Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
0.1 mL
0.3 mL
1 mL
2 mL
3 mL
5 mL
Height (Metric):
32 mm
40 mm
41 mm
46 mm
61 mm
Diameter (Metric) Outer:
12 mm
13 mm
20 mm
Volume (English):
0.027 dr.
0.1 dr.
0.3 dr.
0.5 dr.
0.8 dr.
1.4 dr.
Unit Size:
Pack of 12
Product Code. 12663355
Brand: DWK Life Sciences W986211NG
Ideal for small scale reactions, centrifugation, storage, packaging and shipping of vital samples.
  • Not graduated
  • Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
  • Conical interior bottoms are designed to provide total downward drainage for maximum sample retrieval
Order Info
Closures are sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Inner 0.196 in.
Diameter (Metric) Inner 5 mm
Color Clear
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass (Low Extractable)
Volume (Metric) 0.1 mL
For Use With (Application) Used for small scale reactions, centrifugation, storage, packaging and shipping of vital samples
Graduated No
Style Conical V
Volume (English) 0.027 dr.
Closure Type Crimp Cap
Height (Metric) 32 mm
Certifications/Compliance ASTM E438, USP Type I, Type I, Class A