Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Tear-Off Seals
Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
13 mm
20 mm
30 mm
Unit Size:
Pack of 1000
Description
Tear-off seal can be removed completely allowing content to pour from bottle
- Aluminum
- Unlined
Specifications
Specifications
| Color | Red |
| Material | Aluminum |
| Quantity | 1000/Cs. |
| Type | Complete Tear-off Cap |
| Closure Size | 13 mm |