Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Center Disc Tear-Out Seals
Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Red
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1000
Description
Exposes the top of the rubber stopper to the syringe when the center disc is opened or removed
- Available in 13 or 20mm O.D.
Specifications
Specifications
| Color | Red |
| Material | Aluminum |
| Quantity | 1000/Cs. |
| Closure Size | 13 mm |
| Product Type | Center Disc Tear-out Cap |