Learn More
DWK Life Sciences Wheaton™ Straight Plug Rubber Stoppers
Product Code. 11734199 Shop All Wheaton Science Products Products
Click to view available options
Color:
Gray
Red
Diameter (Metric) Outer:
13 mm
20 mm
Material:
Chlorobutyl-Isoprene
FEP
Rubber
Product Type:
Straight Plug Stopper
Unit Size:
Pack of 1000
Product Code. 11734199
Brand: DWK Life Sciences 224100070
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides maximum sealing for vials with straight-wall glass finishes. Wheaton™ Straight Plug Rubber Stoppers are manufactured from the highest quality raw materials.
  • Autoclavable
  • Available in a variety of sizes/colors
  • Manufactured to yield exceptional dimensional stability
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vacule Ampules
Color Red
Diameter (Metric) Outer 13 mm
Material Rubber
Product Type Straight Plug Stopper