Learn More
DWK Life Sciences Wheaton™ Stainless-Steel Shaft Assembly Kit For Celstir
Product Code. p-3303350 Shop All Wheaton Science Products Products
Click to view available options
For Use With (Equipment):
1000mL Flask
125mL Flask
250mL Flask
25mL Flask
6000mL Flask
8000mL Flask
Unit Size:
Each
Product Code. 11551295
Brand: DWK Life Sciences 356880
Adjustable paddle blade impeller for better mixing
  • Provide maximum surface interface between culture and flask atmosphere
  • Ideal for microcarrier and suspension cultures such as insect cells, hybridomas, and adapted cell lines
  • Made from Wheaton 33 low-extractable borosilicate glass that conforms to ASTM™ Type I, Class A and USP Type I requirements
  • Impeller rod does not protrude up through top cap
  • Sidearms for 500mL to 8L flask allow vented caps which enables the flask to breathe while protecting from contamination
  • 1:1 headspace for good gas exchange
  • No contamination from stirring rod
  • Bottom dimple on 125mL and larger flask improves circulation and reduces accumulation of cells in center of the flask
  • Sizes 125 to 1000mL are graduated in 50mL increments; sizes 3000 to 8000mL are graduated in 500mL increments; sizes 25mL and 50mL are non-graduated
  • Completely autoclavable at 121°C for 20 minutes
Order Info
Assemblies 500mL and larger can be used with the vented cap (Cat. No. 22-288-526)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 250mL Flask
Type Shaft Assembly Kit