Learn More
DWK Life Sciences Wheaton™ Serum Vials
Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
3 mL
5 mL
10 mL
10.0 mL
20 mL
Color:
Amber
Clear
Diameter (Metric) Outer:
15 mm
17 mm
22 mm
24 mm
30.5 mm
Unit Size:
Pack of 120
Pack of 144
Beskrivning
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
- Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
- Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
- Withstand extreme temperature variations
- Necks finished for aluminum crimp seals
- Blowback feature of neck and specially designed bottom radius for lyophilization applications
- Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Uppfyllande
Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards
Specifikationer
Specifikationer
| Diameter (English) Inner | 0.27 in. |
| Diameter (Metric) Inner | 7 mm |
| Color | Clear |
| Diameter (Metric) Outer | 17 mm |
| Material | Borosilicate Glass |
| Volume (Metric) | 3 mL |
| For Use With (Application) | For lyophilization, packaging, sampling and freeze drying applications |
| Volume (English) | 0.8 dr. |
| Autoclavable | Autoclavable |
| Closure Type | Crimp Cap |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering