Learn More
DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
Alle ansehen Wheaton Science Products Produkte
missing translation for 'orderingAttributeHoverText'
Septum:
PTFE/Silicone
Color (Seal):
White/Transparent Blue
missing translation for 'unitSize'
Pack of 100
Description
- For use with CTC Analytics Systems requiring magnetic closures
- Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
Specifications
Specifications
| Color (Seal) | White/Transparent Blue |
| Material | PTFE, Silicone |
| Septum | PTFE/Silicone |
| Type | Screw Cap |