Learn More
DWK Life Sciences Wheaton™ Arsine Generator With Clear–Seal Joint And 24/40 Standard Taper
Product Code. 12495472 Shop All Wheaton Science Products Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
12495472 24/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12495472 Supplier DWK Life Sciences Supplier No. 180020
  • ONLINE EXCLUSIVE PRICE
    198.45€
    173.15€ / Pack of 1
    Save 25.30€
    12% Off
Estimated Shipment: 10-03-2027
Engineered for arsine analysis using the silver diethyldithiocarbamate (SDDC) colorimetric method
  • Manufactured from WHEATON 33 low extractable borosilicate glass
Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements; Reference: £Standard Test Method for Arsenic in Water,£ASTM D2972
Specifications
Product Type Arsine Generator
Standard Taper Joint 24/40
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For arsine analysis using SDDC method
Includes 125mL erlenmeyer flask scrubber tube, socket joint, size 12 stainless steel ball joint clamp