Learn More
DWK Life Sciences Wheaton™ Arsine Generator, Modified
Product Code. 11722235 Shop All Wheaton Science Products Products
Change view
Click to view available options
Standard Taper Joint:
24/40
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
11722235 24/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11722235 Supplier DWK Life Sciences Supplier No. 180023
  • ONLINE EXCLUSIVE PRICE
    177.45€
    168.10€ / Pack of 1
    Save 9.35€
    5% Off
 Restricted Item
Estimated Shipment: 27-10-2026
Modified for Arsine analysis
  • Modified version of catalog number 180020 for arsine analysis
  • The freestanding system has a standard taper 24/40 Clear-Seal™ joint
  • One piece scrubber/absorber unit
  • Low extractable borosilicate glass
  • Eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes
  • Manufactured from WHEATON 33 low-extractable borosilicate glass
Compliance
Conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Specifications
Product Type Arsine Generator
Standard Taper Joint 24/40
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Application) For arsine analysis using SDDC method
Includes 125mL erlenmeyer flask with 24/40 joint, one-piece scrubber/absorber unit