Learn More
DWK Life Sciences Wheaton™ 12 x 32mm Large Opening E-Z Vial With 11mm Snap Caps
Shop All Wheaton Science Products Products
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
Specifications
Specifications
| Color | Natural |
| Material | Polyethylene |
| Closure Type | Snap Cap |
| Quantity | 1000 Cs. |
| Closure Size | 11 mm |