Learn More
DWK Life Sciences WHEATON™ Glass Staining Dish
Product Code. 10633533 Shop All DWK Life Sciences Products
Change view
Click to view available options
Length (Metric):
118 mm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10633533 118 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10633533 Supplier DWK Life Sciences Supplier No. 900301
  • ONLINE EXCLUSIVE PRICE
    342.00€
    318.09€ / Pack of 3
    Save 23.91€
    6% Off
Estimated Shipment: 23-11-2026
The WHEATON™ Glass Staining Dish is made of soda-lime glass and accommodates 16, 20, and 30 slide racks. It offers a flexible, durable solution for histology and pathology slide staining workflows.
Made of sode-lime glass.
  • Made of soda-lime glass for durability and chemical resistance
  • Accommodates 16, 20, and 30 slide racks
  • Approximate inside dimensions: 118L x 88W x 70mm D
  • Slide rack sold separately
Specifications
Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Material Soda Lime Glass
Type Staining Dish
For Use With (Equipment) Standard microscope slides, Slide Rack (Mfr. No. 900304)
Dimensions (L x W) 4.6 x 3.4 in. (11.8 x 8.8 cm)
Length (English) 4.6 in.
Length (Metric) 118 mm
Width (English) 3.4 in.
Width (Metric) 88 mm
Height (Metric) 66 mm
Depth (English) Inside 2.7 in.
Depth (Metric) 70 mm
Depth (Metric) Inside 7 cm