Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Product Code. p-4368053 Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
20 mL
24 mL
25 mL
30 mL
40 mL
Color:
Amber
Clear
Packaging Type:
2 x Shrink-wrapped Packs
Partitioned Tray
Unit Size:
Pack of 200
Product Code. 10380675
Brand: DWK Life Sciences 224833
Packaged in partitioned trays, without caps
Partitioned trays provide an easy way to inventory samples or to store empty vials.
  • Autoclavable
  • Clear or Amber Vials
    • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
    • Amber vials made from borosilicate glass that protects light-sensitive products
    Order Info
    Caps available separately
    Compliance
    Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
    TRUSTED_SUSTAINABILITY
    Specifications
    Color Clear
    Diameter (Metric) Outer 28 mm
    Material Borosilicate Glass
    Volume (Metric) 40 mL
    Packaging Type Partitioned Tray
    Thread Style Screw
    Volume (English) 10 dr.
    Autoclavable Autoclavable
    Closure Type Screw Cap
    Height (Metric) 95 mm
    Certifications/Compliance ASTM E438, USP Type I, Type I, Class A
    Type Sample Vial in Lab File
    For Use With (Equipment) M-T File™ and Vial Rack