Learn More
DWK Life Sciences Wheaton™ Glass 20mL Scintillation Vials: Polypropylene Caps
Product Code. 10079010 Shop All Wheaton Science Products Products
Click to view available options
Material (Liner):
Foamed Polyethylene
Metal Foil
Closure Size:
12.3 mm
13.1 mm
Unit Size:
Pack of 100
Pack of 500
Product Code. 10079010
Brand: DWK Life Sciences 986541
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Maintain integrity of seal with variety of cap liners. Wheaton Science Products™ Glass 20mL Scintillation Vials: Polypropylene Caps are Chemically inert, suitable for strong solvents and high-purity chemicals.
  • Chemically inert, suitable for strong solvents and high-purity chemicals
  • Variety of cap liners available to maintain integrity of seal
  • Background counts are consistent and low; ultraviolet transmission is high
  • With screw caps attached to vials
Order Info
O.D. x H: 1.125 x 2.375 in. (28 x 61mm) with cap on; vials are packed in individual cells in five trays of 100 per case
Compliance
Meets ASTM™ E438 Type I, and Class A standards.
TRUSTED_SUSTAINABILITY
Specifications
Closure Color White
Material (Liner) Metal Foil
Diameter (English) Outer 1.1 in.
Color Clear
Diameter (Metric) Outer 28 mm
Material Borosilicate Glass
Volume (Metric) 20 mL
For Use With (Application) Good barrier properties and resistance to hydrocarbons, oils, ketones and alcohols
Height (English) 2.4 in.
Packaging Type Cellular Trays
Volume (English) 0.67 oz.
Volume (Metric) Total 20 mL
Closure Material Polypropylene
Closure Type Screw Cap
Height (Metric) 61 mm
Certifications/Compliance ASTM E438, Type I, Class A
Type Scintillation Vial
Closure Size 12.3 mm