Learn More
DWK Life Sciences DURAN™ PURE Premium Screw Caps
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 45
Unit Size:
Pack of 5
Description
- Made from fluoropolymer
- Material contact surfacesmade from PTFE
- Colorant and pigment free
- Available in two DIN Thread sizes: GL 25 and GL 45
Specifications
Specifications
| Description | Premium Screw Cap, DIN Thread Size: GL 45 |
| Material | Fluoropolymer |
| Thread Size (Metric) | GL 45 |