Learn More
DWK Life Sciences Wheaton™ Deck Kit for Compact Roller System for Mini Bottles
Shop All Wheaton Science Products ProductsDescription
- Can add up to a maximum of 2 kits per unit.
Specifications
Specifications
| Product Type | Deck Kit |
| Weight (English) | 9 lb. |
| Weight (Metric) | 4 kg |
| Color | Charcoal |
| For Use With (Equipment) | Mini Bottle Roller Culture Apparatus |
| Includes | Assembled roller deck, 4 support posts, drive belt, necessary hardware and assembly instructions |