Learn More
DWK Life Sciences Wheaton™ Connection Connectors
Shop All Wheaton Science Products Products
Click to view available options
Thread Size:
12-425 and 24-400
20-400 and 24-400
Unit Size:
Pack of 6
Description
- Screw cap configuration creates a grease-free, vacuum- tight seal
- Inert contact surface
- Easy to assemble
- Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
- Available with white polypropylene or black phenolic caps
- Phenolic connectors have a working temperature of up to 180
Specifications
Specifications
| Certifications/Compliance | USP Type I, ASTM E438 Type I, Class A |
| For Use With (Application) | Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere |
| For Use With (Equipment) | Micro soxhlet extraction apparatus |
| Includes | Screw cap |
| Material | Borosilicate Glass, Polypropylene |
| Product Type | Screw Thread Connector |
| Thread Size | 20-400 and 24-400 |