Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
Shop All Wheaton Science Products Products
Click to view available options
Color:
Clear
Volume (Metric):
2 mL
5 mL
10 mL
Unit Size:
Pack of 158
Pack of 200
Pack of 210
Description
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components – a solution for any application
- Complete manufacturing & certificate of analysis traceability
- USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Amber versions for light sensitive content packaging
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Specifications
| Color | Clear |
| Material | Borosilicate Glass (Type 1) |
| Sterility | Sterile |
| Volume (Metric) | 5 mL |
| No. per Case | 210 |
| Product Line | Wheaton CompletePAK |
| Certifications/Compliance | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |