Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
Shop All Wheaton Science Products Products
Click to view available options
Color:
Amber
Clear
Volume (Metric):
2 mL
5 mL
10 mL
missing translation for 'unitSize'
Pack of 104
Pack of 120
Pack of 278
Description
- Sterilized, off-the-shelf, ready-to-use (RtU) products
- Variety of Kitted and stand-alone sterile components – a solution for any application
- Complete manufacturing & certificate of analysis traceability
- Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
- Amber versions for light sensitive content packaging
Certifications
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and lot certified to SAL <10-6, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
Specifications
Specifications
| Color | Clear |
| Material | Borosilicate Glass (Type 1) |
| Sterility | Sterile |
| Volume (Metric) | 10 mL |
| No. per Case | 104/Cs. |
| Product Line | Wheaton CompletePAK |
| Certifications/Compliance | USP 788, 85, and 71, Container Closure Integrity (CCI) certified |