Learn More
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components
Product Code. 12471551 Shop All Wheaton Science Products Products
Click to view available options
Type:
Absorber Top
Cold Finger Condenser
Cold Finger Jacket
Distilling Flask
Inlet Tube
Unit Size:
Each
Product Code. 12471551
Brand: DWK Life Sciences 377163
Only null left
Add to basket
Add to basket
Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)
Compliance
For use according to ASTM D 2036
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cyanide distillation kit
Type Cold Finger Condenser