Få mere at vide
Få mere at vide
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Shop alle Wheaton Science Products produkter
missing translation for 'orderingAttributeHoverText'
Material (Liner):
Linerless
Metal Foil
Polyethylene Disc
Polyseal™
Material:
Polyethylene
Polypropylene
Urea
Closure Size:
15-425
22-400
Förpackningsstorlek
Pack of 1000
Beskrivelse
Screw Caps For Liquid Scintillation Vials
- Screw caps for WHEATON liquid scintillation vials
- Case of 1000 provides an economical alterative
- Not autoclavable
Tekniske data
Tekniske data
| Description | Cap Size: 22-400 |
| Material (Liner) | Polyseal™ |
| Color | White |
| Material | Urea |
| Product Line | Wheaton |
| Autoclavable | Not Autoclavable |
| Quantity | 1000 Cs. |
| Type | Screw Cap |
| Closure Size | 22-400 |
| For Use With (Equipment) | Liquid Scintillation Vial |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion