Learn More
DWK Life Sciences Wheaton™ Aluminum Seals: Flip Cap
Shop All Wheaton Science Products Products
Click to view available options
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1000
Description
Flip Cap Seals are easy to use, autoclavable and tamper-evident
- The red plastic top flips off easily with pressure from the thumb exposing the aluminum seal with center hole and pull ring
- For Vials with a 13mm OD
Specifications
Specifications
| Color | Red |
| Material | Aluminum |
| Quantity | 1000/Cs. |
| Closure Size | 13 mm |
| Product Type | Flip Cap |