Learn More
DWK LIFE SCIENCES™ WHEATON™ 30-Slide Rack
Product Code. 12676756 Shop All DWK Life Sciences Products
Change view
Click to view available options
Slide Capacity:
30
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Slide Capacity Unit Size
12676756 30 Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12676756 Supplier DWK LIFE SCIENCES™ Supplier No. 900234
  • ONLINE EXCLUSIVE PRICE
    255.00€
    198.35€ / Pack of 3
    Save 56.65€
    22% Off
Estimated Shipment: 14-12-2026
The WHEATON™ 30-Slide Rack is constructed from stainless steel, providing durable and corrosion-resistant storage and organisation for up to 30 standard microscope slides in staining and laboratory workflows.
  • Holds up to 30 standard microscope slides
  • Constructed from stainless steel for durability and corrosion resistance
  • Suitable for use in staining and histology laboratory workflows
  • Compatible with standard 3 x 1 in. microscope slides
Specifications
Type Slide Holder
Slide Capacity 30
Closure Type Hinged
Depth (English) 2.75 in.
Depth (Metric) 7 cm
Dimensions (L x W) 4.64 x 3.46 in. (11.8 x 8.8 cm)
For Use With (Equipment) Microscope Slide
Length (English) 4.64 in.
Length (Metric) 11.8 cm
Material Stainless Steel
Width (English) 3.46 in.
Width (Metric) 8.8 cm