Learn More
DWK Life Sciences Wheaton™ 12 x 32mm Glass/Plastic Vials with 0.1mL Limited Volume Insert
Shop All Wheaton Science Products Products
Click to view available options
Color:
Amber
Clear
Closure Type:
Screw Cap
Snap/Crimp
Closure Size:
11 mm
8-425
Unit Size:
Pack of 100
Description
Limited volume, glass/plastic vials
- 0.1mL Limited volume Type 1 borosilicate glass insert
- Vials with glass inner core combine Type I borosilicate glass with thermoplastic polymer outer shell
- Glass is only contact between sample and seal
- Polymer provides excellent dimensional tolerance reliability for capping and instrumentation, and prevents breakage
- Conical interior ensures maximum retrieval of contents without the hassle of using removable inserts
- Screw Top Finish (8-425)
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 12 mm |
| Material | Glass, Plastic |
| Volume (Metric) | 0.1 mL |
| Height (English) | 1.25 in. |
| Includes | 0.1mL glass insert |
| Closure Type | Screw Cap |
| Height (Metric) | 32 mm |
| Quantity | 100 Cs. |
| Closure Size | 8-425 |