Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 58
Product Code. 11305404
Click to view available options
Diameter (Metric):
47 mm
50 mm
100 mm
Unit Size:
Pack of 100
Pack of 25
Pack of 5
Dimensions (L x W):
300 x 600 mm
Product Code. 11305404
Brand: Cytiva 10410319
Only null left
Add to basket
Add to basket
Consists of pure cellulose without any wetting agents
  • Spontaneously wetting, very good wet strength
  • Extremely chemically resistant; suitable for aqueous and organic media
  • Hydrophilic
  • Mechanically stable
  • Sterilizable by all methods
TRUSTED_SUSTAINABILITY
Specifications
Material Regenerated Cellulose
Pore Size 0.2 μm
Hydrophobicity Hydrophilic
Thickness (Metric) 75 μm
Diameter (Metric) 100 mm