Learn More
Cytiva Whatman™ Polyamide Membranes
Product Code. 11358944
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.5 μm
Diameter (Metric):
25 mm
47 mm
50 mm
Unit Size:
Pack of 100
Product Code. 11358944
Brand: Cytiva 10414114
Universal filter made from pure polyamide for clarification and sterile filtration
  • Mechanicall strong
  • Excellent wet/dry strength
  • Hydrophilic for aqueous and organic solutions
  • For up to 135°C
TRUSTED_SUSTAINABILITY
Specifications
Material Polyamide
Pore Size 0.5 μm
Hydrophobicity Hydrophilic
Thickness (Metric) 110 μm
Diameter (Metric) 50 mm