Learn More
Cytiva Whatman™ pH Indicators
Product Code. 11375254
Click to view available options
pH Range:
0 to 14
1 to 11
2 to 9
Quantity:
1 Roll
100 Pack
200 Pack
Unit Size:
Each
Pack of 100
Pack of 200
Product Code. 11375254
Brand: Cytiva 10360005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides rapid determination of pH values. Whatman™ pH Indicators are impregnated with a mixture of several indicators that change color when they come in contact with a particular sample solution.
Determine pH using the supplied color comparison table. Test strips assume a particular color on contact with the sample solution.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use. These pH indicators have been impregnated with a mixture of several indicators
TRUSTED_SUSTAINABILITY
Specifications
pH Range 0 to 14
Format Strip
Includes Color comparison chart
Quantity 200 Pack
Type Indicator Paper