Learn More
Cytiva Whatman™ pH Indicator Papers, CF Strips
Product Code. 11390684
Change view
Click to view available options
pH Range:
0 to 14
4.5 to 10
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
11390684 4.5 to 10 Pack of 100
10377481 0 to 14 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11390684 Supplier Cytiva Supplier No. 2614991
  • ONLINE EXCLUSIVE PRICE
    22.55€
    21.84€ / Pack of 100
    Save 0.71€
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
  • Plastic supports that retain stiffness in liquids
  • Paper segments on each strip have chemically bonded dyes; can't leach into solution
  • Complementary indicator charts provide clear comparison
  • Full-range strips (0 to 14) graduated in 1pH units; wide-range strips (4.5 to 10pH) in 0.5pH units
  • Packs of 100; each strip is approximately 0.25 × 3 in. (6 × 80mm)
TRUSTED_SUSTAINABILITY
Specifications
pH Range 4.5 to 10
Format Strip
Graduations 0.5 pH
Includes Plastic supports, paper segments on each strip, complementary indicator charts
Quantity 100 Pack
Type pH Test Strip, Color Bonded
Dimensions (L x W) 80 x 6 mm
Length (Metric) 80 mm
Width (Metric) 6 mm