Learn More
Cytiva Whatman™ pH Indicator Papers, CF Strips
Product Code. 10377481
Change view
Click to view available options
pH Range:
0 to 14
4.5 to 10
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10377481 0 to 14 Pack of 100
11390684 4.5 to 10 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10377481 Supplier Cytiva Supplier No. 2613991
  • ONLINE EXCLUSIVE PRICE
    27.80€
    22.13€ / Pack of 100
    Save 5.67€
    20% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Provides an extremely clear and accurate visual pH value using four different segments of dye-impregnated indicator papers. Whatman™ pH Indicator Papers, CF Strips use dyes that are chemically bonded to the paper and cannot be leached into solution.
  • Plastic supports that retain stiffness in liquids
  • Paper segments on each strip have chemically bonded dyes; can't leach into solution
  • Complementary indicator charts provide clear comparison
  • Full-range strips (0 to 14) graduated in 1pH units; wide-range strips (4.5 to 10pH) in 0.5pH units
  • Packs of 100; each strip is approximately 0.25 × 3 in. (6 × 80mm)
TRUSTED_SUSTAINABILITY
Specifications
pH Range 0 to 14
Format Strip
Graduations 1 pH
Includes Plastic supports, paper segments on each strip, complementary indicator charts
Quantity 100 Pack
Type pH Test Strip, Color Bonded
Dimensions (L x W) 80 x 6 mm
Length (Metric) 80 mm
Width (Metric) 6 mm