Learn More
Cytiva Whatman™ Nuclepore Hydrophilic Membrane
Product Code. 16675675
missing translation for 'orderingAttributeHoverText'
Pore Size:
1 μm
2 μm
3 μm
8 μm
10 μm
12 μm
Diameter (Metric):
13 mm
19 mm
25 mm
47 mm
90 mm
293 mm
missing translation for 'unitSize'
Pack of 100
Pack of 25
Product Code. 16675675
missing translation for 'mfr': Cytiva 10418701
Only null left
Add to basket
Add to basket
Nuclepore track-etched hydrophilic membranes with highly defined pore size cutoff in sub-micron range
  • Track-etched for highly defined pore size and high pore density
  • Treated with PVP to make it hydrophilic
  • Low extractables minimizing filtration sample contamination risk
  • High chemical resistance
  • Good thermal stability
  • Low, consistent ash
  • Low tare weights
  • Smooth, flat membrane surface for high particle visibility
Applications:
  • Bioassays
  • Cell biology
  • Environmental analysis
  • Microscopy
  • Cytology
  • Parasitology
  • Certain air and oceanography analyses
TRUSTED_SUSTAINABILITY
Specifications
Material Polycarbonate
Pore Size 1 μm
For Use With (Application) Bioassays, Cell biology, Environmental analysis, Microscopy, Cytology, Parasitology, Certain air and oceanography analyses
Quantity 100 Pk.
Type Nuclepore hydrophilic membrane
Diameter (Metric) 13 mm